Anti-NUDT3 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126962-50EA 946 EUR
ABCAAB126962-50
Anti-NUDT3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: NUDT3 (nudix (nucleoside diphosphate linked moiety X)-type motif 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR