Anti-DDX21 Rabbit polyclonal antibody unconjugated

Toimittaja: Abcam
ABCAAB126968-50EA 997 EUR
ABCAAB126968-50
Anti-DDX21 Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: DDX21 (DEAD (Asp-Glu-Ala-Asp) box helicase 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR