Anti-CSRP2BP Rabbit polyclonal antibody unconjugated
Toimittaja: Abcam
ABCAAB127040-50EA 907 EUR
ABCAAB127040-50
Anti-CSRP2BP Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: CSRP2BP (CSRP2 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human