Anti-RBM4B Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB127050-50EA 907 EUR
ABCAAB127050-50
Anti-RBM4B Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RBM4B (RNA binding motif protein 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR