Anti-ZBTB20 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB127702-50EA 997 EUR
ABCAAB127702-50
Anti-ZBTB20 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ZBTB20 (zinc finger and BTB domain containing 20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR