Anti-ZNF334 Rabbit Polyclonal Antibody
Toimittaja: Abcam
ABCAAB127712-50EA 946 EUR
ABCAAB127712-50
Anti-ZNF334 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ZNF334 (zinc finger protein 334)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human