Anti-SERPINB7 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB127752-50EA 946 EUR
ABCAAB127752-50
Anti-SERPINB7 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SERPINB7 (serpin peptidase inhibitor, clade B (ovalbumin), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR