Anti-LRRC59 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB127912-50EA 946 EUR
ABCAAB127912-50
Anti-LRRC59 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: LRRC59 (leucine rich repeat containing 59)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR