Anti-GAP43 Rabbit polyclonal antibody unconjugated

Toimittaja: Abcam
ABCAAB128005-50EA 959 EUR
ABCAAB128005-50
Anti-GAP43 Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: GAP43 (growth associated protein 43)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR