Anti-PGM3 Mouse monoclonal antibody [clone: 1A11]

Toimittaja: Abcam
ABCAAB128094-100EA 928 EUR
ABCAAB128094-100
Anti-PGM3 Mouse monoclonal antibody [clone: 1A11]
Type: Primary
Antigen: PGM3 (phosphoglucomutase 3)
Clonality: monoclonal
Clone: 1A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR