Anti-MAPK1/3 Rabbit Polyclonal Antibody
Toimittaja: Abcam
ABCAAB128159-100EA 997 EUR
ABCAAB128159-100
Anti-MAPK1/3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human