Anti-RRAGA Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB128196-100EA 946 EUR
ABCAAB128196-100
Anti-RRAGA Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RRAGA (Ras-related GTP binding A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR