Anti-RNF5 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB128200-100EA 997 EUR
ABCAAB128200-100
Anti-RNF5 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR