Anti-VIM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: RV202]

Toimittaja: Abcam
ABCAAB128507-500EA 1050 EUR
ABCAAB128507-500
Anti-VIM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: RV202]
Vasta-aineet
Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: RV202
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR