Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 650) [clone: OC2-4E8]

Toimittaja: Novus Biologicals
NOVUNBP1-18971CEA 656 EUR
NOVUNBP1-18971C
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 650) [clone: OC2-4E8]
Vasta-aineet
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-4E8
Conjugation: DyLight® 650
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR