Anti-TLR4 Rabbit Polyclonal Antibody (DyLight® 550)

Toimittaja: Novus Biologicals
NOVUNBP1-78427REA 709 EUR
NOVUNBP1-78427R
Anti-TLR4 Rabbit Polyclonal Antibody (DyLight® 550)
Vasta-aineet
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR