Anti-ABL1 Rabbit Polyclonal Antibody

PK1013-50EA 0 EUR
CALBPK1013-50
Anti-ABL1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: ABL1 (c-abl oncogene 1, non-receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR