Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)
Toimittaja: Rockland Immunochemicals
ROCK610-1303EA 366 EUR
ROCK610-1303
Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)
Vasta-aineet
Anti-Mouse IgG F(c) fragment is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Mouse