Anti-IgA Rabbit Polyclonal Antibody
Toimittaja: Rockland Immunochemicals
ROCK610-4106EA 159 EUR
ROCK610-4106
Anti-IgA Rabbit Polyclonal Antibody
Secondary Rabbit Anti-IgA (alpha chain) Reacts with Mouse
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgA
Reactivity: Mouse