Anti-CDC25A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: US Biological
USBI033576-APEA 852 EUR
USBI033576-AP
Anti-CDC25A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR