Anti-GRB7 Rabbit Polyclonal Antibody (Biotin)

Toimittaja: US Biological
USBI036282-BIOTINEA 852 EUR
USBI036282-BIOTIN
Anti-GRB7 Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: GRB7 (growth factor receptor-bound protein 7)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR