Anti-HMBOX1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Toimittaja: US Biological
USBI036630-FITCEA 852 EUR
USBI036630-FITC
Anti-HMBOX1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HMBOX1 (homeobox containing 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse