Anti-HMFG Rabbit Polyclonal Antibody

Toimittaja: US Biological
USBI036632EA 587 EUR
USBI036632
Anti-HMFG Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Western Blot: 1:1000; Flow Cytometry: 1:10-50

Type: Primary
Antigen: HMFG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR