Anti-MTIF2 Rabbit Polyclonal Antibody (Biotin)
Toimittaja: US Biological
USBI038659-BIOTINEA 852 EUR
USBI038659-BIOTIN
Anti-MTIF2 Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MTIF2 (mitochondrial translational initiation factor 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human