Anti-MTL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: US Biological
USBI038661-HRPEA 852 EUR
USBI038661-HRP
Anti-MTL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MTL5 (metallothionein-like 5, testis-specific (tesmin))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human