Anti-MTMR6 Rabbit Polyclonal Antibody (Biotin)
Toimittaja: US Biological
USBI038669-BIOTINEA 852 EUR
USBI038669-BIOTIN
Anti-MTMR6 Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MTMR6 (myotubularin related protein 6)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human