Anti-MTMR6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Toimittaja: US Biological
USBI038669-FITCEA 852 EUR
USBI038669-FITC
Anti-MTMR6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTMR6 (myotubularin related protein 6)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human