Anti-MTMRF Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: US Biological
USBI038671-APEA 852 EUR
USBI038671-AP
Anti-MTMRF Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MTMRF
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human