Anti-MTMRF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Toimittaja: US Biological
USBI038671-FITCEA 852 EUR
USBI038671-FITC
Anti-MTMRF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MTMRF
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human