Anti-MTNR1A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: US Biological
USBI038672-APEA 678 EUR
USBI038672-AP
Anti-MTNR1A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MTNR1A (melatonin receptor 1A)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human