Anti-NDUFA10 Rabbit Polyclonal Antibody

Toimittaja: US Biological
USBI038856EA 587 EUR
USBI038856
Anti-NDUFA10 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: NDUFA10 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR