Anti-NDUFAF2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Toimittaja: US Biological
USBI038862-FITCEA 852 EUR
USBI038862-FITC
Anti-NDUFAF2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: NDUFAF2 (NADH dehydrogenase (ubiquinone) complex I, assembly factor 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human