Anti-NDUFAF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: US Biological
USBI038862-HRPEA 852 EUR
USBI038862-HRP
Anti-NDUFAF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: NDUFAF2 (NADH dehydrogenase (ubiquinone) complex I, assembly factor 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR