Anti-NDUFC2 Rabbit Polyclonal Antibody

Toimittaja: US Biological
USBI038867EA 587 EUR
USBI038867
Anti-NDUFC2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: NDUFC2 (NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR