Anti-NDUFC2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Toimittaja: US Biological
USBI038867-FITCEA 852 EUR
USBI038867-FITC
Anti-NDUFC2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NDUFC2 (NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR