Anti-NDUFC2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
USBI038867-PEEA 852 EUR
USBI038867-PE
Anti-NDUFC2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NDUFC2 (NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR