Anti-NDUFS6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Toimittaja: US Biological
USBI038870-FITCEA 852 EUR
USBI038870-FITC
Anti-NDUFS6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NDUFS6 (NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR