Anti-SFTPD Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Toimittaja: United States Biological
USBI041643-PEEA 852 EUR
USBI041643-PE
Anti-SFTPD Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SFTPD (surfactant protein D)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human