Anti-SFXN3 Rabbit Polyclonal Antibody (Biotin)
Toimittaja: United States Biological
USBI041645-BIOTINEA 852 EUR
USBI041645-BIOTIN
Anti-SFXN3 Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SFXN3 (sideroflexin 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human