Anti-SGCA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: United States Biological
USBI041647-HRPEA 852 EUR
USBI041647-HRP
Anti-SGCA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SGCA (sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR