Anti-SHANK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Toimittaja: United States Biological
USBI041698-PEEA 852 EUR
USBI041698-PE
Anti-SHANK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SHANK2 (SH3 and multiple ankyrin repeat domains 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse