Anti-SHARPIN Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: United States Biological
USBI041699-APEA 852 EUR
USBI041699-AP
Anti-SHARPIN Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SHARPIN (SHANK-associated RH domain interactor)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR