Anti-SHBG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: United States Biological
USBI041701-APEA 852 EUR
USBI041701-AP
Anti-SHBG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SHBG (sex hormone-binding globulin)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human