Anti-SHC4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Toimittaja: United States Biological
USBI041704-APCEA 852 EUR
USBI041704-APC
Anti-SHC4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SHC4 (SHC (Src homology 2 domain containing) family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse