Anti-SHC4 Rabbit Polyclonal Antibody (Biotin)

Toimittaja: United States Biological
USBI041705-BIOTINEA 852 EUR
USBI041705-BIOTIN
Anti-SHC4 Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: SHC4 (SHC (Src homology 2 domain containing) family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR