Anti-SHOC2 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBI041717EA 587 EUR
USBI041717
Anti-SHOC2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SHOC2 (soc-2 suppressor of clear homolog (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR