Anti-SHOC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Toimittaja: United States Biological
USBI041717-APCEA 852 EUR
USBI041717-APC
Anti-SHOC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SHOC2 (soc-2 suppressor of clear homolog (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR