Anti-UTS2R Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Toimittaja: United States Biological
USBI043691-APCEA 852 EUR
USBI043691-APC
Anti-UTS2R Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: UTS2R (urotensin 2 receptor)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human