Anti-UTS2R Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Toimittaja: United States Biological
USBI043691-FITCEA 852 EUR
USBI043691-FITC
Anti-UTS2R Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: UTS2R (urotensin 2 receptor)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human