Anti-UTS2R Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Toimittaja: United States Biological
USBI043691-PEEA 852 EUR
USBI043691-PE
Anti-UTS2R Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: UTS2R (urotensin 2 receptor)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human